APOH039FD1B1 Apollo Scientific Ltd

2,2,4-Trimethyl-1,4,2-oxazasilinane

  • MDL number MFCD09263699
  • Cas Number : 10196-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=38989

Safety Information

signalword Danger
pcodes P210;P280;P301;P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=38989
pictograms GHS02;GHS05;GHS07
hstatements H226;H302;H312;H332;H314;H335
Availability details for
Quantity
  • Availability
  • Add to Cart