APOH039FD1B5 Apollo Scientific Ltd

2-Bromo-3�bromopropiophenone

  • MDL number MFCD04118000
  • Cas Number : 76650-08-3
  • Molecular Weight : 291.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-bromopropiophenone

Safety Information

signalword Danger
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-3-bromopropiophenone
pictograms GHS05;GHS07
hstatements H302;H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart