APOH039FD1B9 Apollo Scientific Ltd

(3-Bromophenyl)methanethiol

  • MDL number MFCD06798010
  • Cas Number : 886497-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Flash Point 120
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=74714

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=74714
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart