APOH039FD1BD Apollo Scientific Ltd

4-Bromo-2-nitrothiophenol

  • MDL number MFCD14652161
  • Cas Number : 76209-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Bromo-2-nitrobenzenethiol
Storage temp. Store at 2-8
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=109868

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=109868
Availability details for
Quantity
  • Availability
  • Add to Cart