APOH039FD1BE Apollo Scientific Ltd

3-Bromo-4-nitrothiophenol

  • MDL number MFCD22481215
  • Cas Number : 1823075-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Bromo-4-nitrobenzenethiol
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=109869

Safety Information

signalword Warning
pcodes P260;P280;P301;P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=109869
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart