APOH039FD1C0 Apollo Scientific Ltd

tert-Butyl 1,2,3-oxathiazinane-3-carboxylate 2,2-dioxide

  • MDL number MFCD22123271
  • Cas Number : 521267-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 2,2-dioxooxathiazinane-3-carboxylate
Storage temp. Store at 2-8
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=111096

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=111096
Availability details for
Quantity
  • Availability
  • Add to Cart