APOH039FD1C3 Apollo Scientific Ltd

3-Bromo-2,4,5-trifluorobenzoic acid

  • MDL number MFCD05664065
  • Cas Number : 104222-42-6
  • Molecular Weight : 254.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 122-123
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-245-trifluorobenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-245-trifluorobenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart