APOH039FD1C9 Apollo Scientific Ltd

2-Bromo-3',4'-difluoropropiophenone

  • MDL number MFCD11182355
  • Cas Number : 23384-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=109159

Safety Information

signalword Danger
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=109159
pictograms GHS05;GHS07
hstatements H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart