APOH039FD1CC Apollo Scientific Ltd

4-bromo-3-fluorophenylthiocyanate

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=106909

Safety Information

signalword Danger
pcodes P260;P280;P301;P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=106909
pictograms GHS05;GHS07;GHS08
hstatements H302;H312;H332;H314;H317;H334
Availability details for
Quantity
  • Availability
  • Add to Cart