APOH039FD1CD Apollo Scientific Ltd

2-Bromo-3’-(trifluoromethyl)propiophenone

  • MDL number MFCD18392396
  • Cas Number : 38786-68-4
  • Molecular Weight : 281.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-1-[3-(trifluoromethyl)phenyl]propan-1-one
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-3-trifluoromethylpropiophenone

Safety Information

signalword Warning
pcodes P280;P261;P271
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart