APOH039FD1CF Apollo Scientific Ltd

3-bromo-4-fluorophenylthiocyanate

  • MDL number MFCD30723328
  • Cas Number : 2149601-24-9
  • Molecular Weight : 232.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-1-fluoro-4-thiocyanatobenzene
Inchi Code 1S/C7H3BrFNS/c8-6-3-5(11-4-10)1-2-7(6)9/h1-3H
Inchi Key UFQCMTVEHGLNNP-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-4-fluorophenylthiocyanate

Safety Information

signalword Danger
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromo-4-fluorophenylthiocyanate
pictograms GHS05;GHS07;GHS08
hstatements H302;H314;H334
Availability details for
Quantity
  • Availability
  • Add to Cart