APOH039FD1DC Apollo Scientific Ltd

3-Fluoro-2-nitrothiophenol

  • MDL number MFCD22481300
  • Cas Number : 1823014-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-Fluoro-2-nitrobenzenethiol
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=109870

Safety Information

signalword Warning
pcodes P260;P280;P301;P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=109870
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart