APOH039FD1DE Apollo Scientific Ltd

2,2,2,2',6'-Pentafluoro-3'-(trifluoromethyl)acetophenone

  • Cas Number : 2149589-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-[2,6-Difluoro-3-(trifluoromethyl)phenyl]-2,2,2-trifluoroethanone
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=110629

Safety Information

signalword Warning
pcodes P260;P280;P301;P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=110629
pictograms GHS07
hstatements H302;H312;H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart