APOH039FD1E3 Apollo Scientific Ltd

2-Bromo-4'-(trifluoromethyl)propiophenone

  • MDL number MFCD17010234
  • Cas Number : 95728-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-1-[4-(trifluoromethyl)phenyl]propan-1-one, 4-(2-Bromopropanoyl)benzotrifluoride
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=47773

Safety Information

signalword Danger
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=47773
pictograms GHS05;GHS07
hstatements H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart