APOH03CD8DDA Apollo Scientific Ltd

2,3-Difluoro-4-(trifluoromethoxy)benzaldehyde

  • Cas Number : 2244086-99-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=113421

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=113421
Availability details for
Quantity
  • Availability
  • Add to Cart