APOH03CD8E23 Apollo Scientific Ltd

2-Bromo-6-chlorostyrene

  • MDL number MFCD31561244

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=113686

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=113686
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart