APOH03CD8E7B Apollo Scientific Ltd

2-(2,2-Dibromovinyl)-4-chloro-1-fluorobenzene

  • MDL number MFCD31561229
  • Cas Number : 1881272-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=113934

Safety Information

signalword Danger
pcodes P260;P280;P301+P310
MSDS http://www.apolloscientific.co.uk/display_item.php?id=113934
pictograms GHS05;GHS06
hstatements H301;H312+H332;H318
Availability details for
Quantity
  • Availability
  • Add to Cart