APOH03CD8EAD Apollo Scientific Ltd

1-(Bromomethyl)-3-nitro-5-(trifluoromethyl)benzene 250mg pack

  • MDL number MFCD11044988
  • Cas Number : 180146-67-2
  • Molecular Weight : 284.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-bromomethyl-3-nitro-5-trifluoromethylbenzene

Safety Information

signalword Danger
pcodes P260;P271;P280
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart