APOH03CD8EC7 Apollo Scientific Ltd

Methyl 2-Bromo-6-fluorobenzoate

  • MDL number MFCD10000922
  • Cas Number : 820236-81-5
  • Molecular Weight : 233.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-bromo-6-fluorobenzoate
Inchi Code 1S/C8H6BrFO2/c1-12-8(11)7-5(9)3-2-4-6(7)10/h2-4H,1H3
Inchi Key TZQFXIYFLBBLQJ-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/methyl-2-bromo-6-fluorobenzoate

Safety Information

signalword Warning
pcodes P280;P302+P352;P304+P340
MSDS https://store.apolloscientific.co.uk/product/methyl-2-bromo-6-fluorobenzoate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart