APOH03CD8F0E Apollo Scientific Ltd

2-Bromo-6-nitrobenzyl alcohol

  • MDL number MFCD11036298
  • Cas Number : 861106-91-4
  • Molecular Weight : 232.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-bromo-6-nitrophenyl)methanol
Inchi Code 1S/C7H6BrNO3/c8-6-2-1-3-7(9(11)12)5(6)4-10/h1-3,10H,4H2
Inchi Key QLLXWADSIGOEQZ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-nitrobenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-nitrobenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart