APOH03CD8F27 Apollo Scientific Ltd

Methyl 4,5,6,7-tetrahydrothiazolo[4,5-c]pyridine-2-carboxylate

  • Cas Number : 1256806-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=113874

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=113874
Availability details for
Quantity
  • Availability
  • Add to Cart