APOH03CD8F29 Apollo Scientific Ltd

tert-Butyl 2-amino-6,7-dihydrothiazolo[4,5-c]pyridine-5(4H)-carboxylate

  • MDL number MFCD11045435
  • Cas Number : 1002355-91-0
  • Molecular Weight : 255.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 2-amino-6,7-dihydro[1,3]thiazolo[4,5-c]pyridine-5(4H)-carboxylate
Inchi Code 1S/C11H17N3O2S/c1-11(2,3)16-10(15)14-5-4-8-7(6-14)13-9(12)17-8/h4-6H2,1-3H3,(H2,12,13)
Inchi Key UPBCRQUYNFQJFR-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/tert-butyl-2-amino-67-dihydrothiazolo45-cpyridine-54h-carboxylate

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS https://store.apolloscientific.co.uk/product/tert-butyl-2-amino-67-dihydrothiazolo45-cpyridine-54h-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart