APOH03CD8F2C Apollo Scientific Ltd

2-Bromo-4,5,6,7-tetrahydro-5-methylthiazolo[4,5-c]pyridine

  • MDL number MFCD22571362
  • Cas Number : 1280704-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=113880

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS http://www.apolloscientific.co.uk/display_item.php?id=113880
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart