APOH03CD8F2E Apollo Scientific Ltd

4-Methyl-2-phenylthiazol-5-ylboronic acid, pinacol ester

  • Cas Number : 690632-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Methyl-2-phenyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-thiazole
Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=113882

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=113882
Availability details for
Quantity
  • Availability
  • Add to Cart