APOH03CD8F40 Apollo Scientific Ltd

7-Bromo-2-mercaptoquinazolin-4(3H)-one

  • MDL number MFCD31561260
  • Cas Number : 1463501-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=113901

Safety Information

signalword Warning
pcodes P260;P280;P301+P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=113901
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart