APOH03CD8F49 Apollo Scientific Ltd

1-(Quinolin-8-yl)ethanone

  • MDL number MFCD00047620
  • Cas Number : 56234-20-9
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 42-45
Iupac Name 1-(8-quinolinyl)ethanone
Inchi Code 1S/C11H9NO/c1-8(13)10-6-2-4-9-5-3-7-12-11(9)10/h2-7H,1H3
Inchi Key SHVCKCOMBWIPBE-UHFFFAOYSA-N

Description

Flash Point 176/12mm
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 42-45
Product Link https://store.apolloscientific.co.uk/product/1-quinolin-8-ylethanone

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/1-quinolin-8-ylethanone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart