APOH03CD8F4D Apollo Scientific Ltd

4,5,6,7-Tetrahydro-5-methylthiazolo[5,4-c]pyridin-2-amine

  • MDL number MFCD02854749
  • Cas Number : 17899-48-8
  • Molecular Weight : 169.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4567-tetrahydro-5-methylthiazolo54-cpyridin-2-amine

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS https://store.apolloscientific.co.uk/product/4567-tetrahydro-5-methylthiazolo54-cpyridin-2-amine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart