APOH03CD8F57 Apollo Scientific Ltd

3-Bromo-4-fluoro-2-nitrophenol

  • MDL number MFCD23132693
  • Cas Number : 1374772-55-0
  • Molecular Weight : 236

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-4-fluoro-2-nitrophenol
Inchi Code 1S/C6H3BrFNO3/c7-5-3(8)1-2-4(10)6(5)9(11)12/h1-2,10H
Inchi Key NRHXZOPTZSJCQL-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromo-4-fluoro-2-nitrophenol

Safety Information

signalword Warning
pcodes P301+P312;P302+P352;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/3-bromo-4-fluoro-2-nitrophenol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart