APOH03CD8F5B Apollo Scientific Ltd

3-Cyclopropylpyrazin-2-amine

  • MDL number MFCD19215706
  • Cas Number : 1190969-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=113936

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=113936
pictograms GHS05;GHS07
hstatements H302;H315;H335;H319
Availability details for
Quantity
  • Availability
  • Add to Cart