APOH03CD8F63 Apollo Scientific Ltd

5-Bromo-3-phenylpyrazin-2-amine

  • MDL number MFCD12547094
  • Cas Number : 344940-70-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-bromo-3-phenylpyrazin-2-amine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-bromo-3-phenylpyrazin-2-amine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart