APOH03CD8F71 Apollo Scientific Ltd

6-Bromo-8-chloroimidazo[1,2-a]pyrazine

  • MDL number MFCD11846485
  • Cas Number : 1208083-37-7
  • Molecular Weight : 232.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-8-chloroimidazo[1,2-a]pyrazine
Inchi Code 1S/C6H3BrClN3/c7-4-3-11-2-1-9-6(11)5(8)10-4/h1-3H
Inchi Key QIRIACFCTBDGFD-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-bromo-8-chloroimidazo12-apyrazine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/6-bromo-8-chloroimidazo12-apyrazine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart