APOH03CD8F73 Apollo Scientific Ltd

3-Chloro-5-iodopyrazin-2-amine

  • MDL number MFCD14706048
  • Cas Number : 1252597-70-8
  • Molecular Weight : 255.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-chloro-5-iodo-2-pyrazinamine
Inchi Code 1S/C4H3ClIN3/c5-3-4(7)8-1-2(6)9-3/h1H,(H2,7,8)
Inchi Key KVGKDDCJSHJYCS-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-5-iodopyrazin-2-amine

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-chloro-5-iodopyrazin-2-amine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart