APOH03CD9AFA Apollo Scientific Ltd

8-Aminoquinoline-7-carboxaldehyde

  • MDL number MFCD11110554
  • Cas Number : 158753-17-4
  • Molecular Weight : 172.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 85-86
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/8-aminoquinoline-7-carboxaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/8-aminoquinoline-7-carboxaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart