APOH03CD9AFB Apollo Scientific Ltd

2-Bromo-6-nitrobenzoic acid

  • MDL number MFCD13688997
  • Cas Number : 38876-67-4
  • Molecular Weight : 246.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177-178
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-nitrobenzoic-acid

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-nitrobenzoic-acid
pictograms GHS07
hstatements H302+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart