APOH03CD9AFC Apollo Scientific Ltd

2-Bromo-6-nitrobenzyl bromide

  • MDL number MFCD11036286
  • Cas Number : 58579-54-7
  • Molecular Weight : 294.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-bromo-2-(bromomethyl)-3-nitrobenzene
Inchi Code 1S/C7H5Br2NO2/c8-4-5-6(9)2-1-3-7(5)10(11)12/h1-3H,4H2
Inchi Key JCBTXEUYUWGIMU-UHFFFAOYSA-N

Description

Synonym 1-Bromo-2-(bromomethyl)-3-nitrobenzene
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-nitrobenzyl-bromide

Safety Information

signalword Danger
pcodes P280;P302+P352;P261
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-nitrobenzyl-bromide
pictograms GHS05;GHS07
hstatements H315;H318;H335
Availability details for
Quantity
  • Availability
  • Add to Cart