APOH03CD9AFD Apollo Scientific Ltd

1,3-Dimethyl-4-nitro-1H-pyrazole

  • MDL number MFCD00464263
  • Cas Number : 3920-38-5
  • Molecular Weight : 141.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1,3-dimethyl-4-nitro-1H-pyrazole
Inchi Code 1S/C5H7N3O2/c1-4-5(8(9)10)3-7(2)6-4/h3H,1-2H3
Inchi Key AKFKERAVXIGUNB-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/13-dimethyl-4-nitro-1h-pyrazole

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/13-dimethyl-4-nitro-1h-pyrazole
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart