APOH03CD9B08 Apollo Scientific Ltd

Fmoc-Cys(Me)-OH

  • MDL number MFCD00237014
  • Cas Number : 138021-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-methylsulfanylpropanoic acid
Storage temp. Ambient temperature
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=112121

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=112121
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart