APOH03CD9B0F Apollo Scientific Ltd

5-(Difluoromethyl)pyridine-2-carboxylic acid

  • MDL number MFCD18256209
  • Cas Number : 859538-41-3
  • Molecular Weight : 173.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-(difluoromethyl)-2-pyridinecarboxylic acid
Inchi Code 1S/C7H5F2NO2/c8-6(9)4-1-2-5(7(11)12)10-3-4/h1-3,6H,(H,11,12)
Inchi Key MKLFNFRBDNMKIU-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-difluoromethylpyridine-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/5-difluoromethylpyridine-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart