APOH03CD9B11 Apollo Scientific Ltd

3-Iodo-5-(trifluoromethyl)benzonitrile

  • MDL number MFCD12922961
  • Cas Number : 691877-05-1
  • Molecular Weight : 297.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-iodo-5-(trifluoromethyl)benzonitrile
Inchi Code 1S/C8H3F3IN/c9-8(10,11)6-1-5(4-13)2-7(12)3-6/h1-3H
Inchi Key PZPKHYOVFCEKBO-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-iodo-5-trifluoromethylbenzonitrile

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-iodo-5-trifluoromethylbenzonitrile
pictograms GHS06
hstatements H301;H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart