APOH03CD9B18 Apollo Scientific Ltd

4-(Difluoromethoxy)-3-hydroxybenzoic acid

  • MDL number MFCD24497448
  • Cas Number : 913985-07-6
  • Molecular Weight : 204.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-difluoromethoxy-3-hydroxybenzoic-acid

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-difluoromethoxy-3-hydroxybenzoic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart