APOH04239DF9 Apollo Scientific Ltd

3-Chloro-5-nitrobenzyl alcohol

  • MDL number MFCD08236779
  • Cas Number : 79944-62-0
  • Molecular Weight : 187.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (3-chloro-5-nitrophenyl)methanol
Inchi Code 1S/C7H6ClNO3/c8-6-1-5(4-10)2-7(3-6)9(11)12/h1-3,10H,4H2
Inchi Key LKBQUSJJIYIBDP-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-chloro-5-nitrobenzyl-alcohol

Safety Information

signalword Warning
pcodes P280;P302+P352
MSDS https://store.apolloscientific.co.uk/product/3-chloro-5-nitrobenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart