APOH04239E1F Apollo Scientific Ltd

3-Iodo-5-nitrobenzyl bromide

  • Cas Number : 1261644-60-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. TBC
Shipping Temperature TBC
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=115093

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=115093
Availability details for
Quantity
  • Availability
  • Add to Cart