APOH04239EB6 Apollo Scientific Ltd

2-(Aminomethyl)-3-bromopyridine hydrochloride

  • MDL number MFCD20923742
  • Cas Number : 1052271-58-5
  • Molecular Weight : 223.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-aminomethyl-3-bromopyridine-hydrochloride

Safety Information

signalword Warning
pcodes P260;P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/2-aminomethyl-3-bromopyridine-hydrochloride
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart