APOH04239EC9 Apollo Scientific Ltd

7-Fluorobenzo[b]thiophene

  • MDL number MFCD17169311
  • Cas Number : 346592-74-3
  • Molecular Weight : 152.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/7-fluorobenzobthiophene

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/7-fluorobenzobthiophene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart