APOH0430C0B4 Apollo Scientific Ltd

4-Bromo-1H-pyrazole-5-carboxaldehyde dimer

  • MDL number MFCD08445454
  • Cas Number : 1212466-82-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-180
Synonym 3,8-Dibromo-4H,9H-dipyrazolo[1,5-a:1',5'-d]pyrazine-4,9-diol, 3,8-Dibromo-4,9-dihydroxy-4H,9H-dipyrazolo[1,5-a:1',5'-d]pyrazine
Purity 95
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13431

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13431
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart