APOH0430C0B7 Apollo Scientific Ltd

Cinnamaldoxime

  • MDL number MFCD00019969
  • Cas Number : 13372-81-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-125
Synonym 3-Phenylacrylaldehyde oxime, Cinnamaldehyde oxime
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=13962

Safety Information

signalword Danger
pcodes P280;P260;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=13962
pictograms GHS06
hstatements H301
Availability details for
Quantity
  • Availability
  • Add to Cart