APOH0430C0B9 Apollo Scientific Ltd

Deoxy-bigchap

  • MDL number MFCD00161482
  • Cas Number : 86303-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym N,N-Bis[3-(D-gluconamido)propyl]deoxycholamide
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=14075

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=14075
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart