APOH0430C0BC Apollo Scientific Ltd

trans-2,4-Dichlorocinnamic acid

  • MDL number MFCD00004373
  • Cas Number : 20595-45-3
  • Molecular Weight : 217.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 233-235
Iupac Name (2E)-3-(2,4-dichlorophenyl)-2-propenoic acid
Inchi Code 1S/C9H6Cl2O2/c10-7-3-1-6(8(11)5-7)2-4-9(12)13/h1-5H,(H,12,13)/b4-2+
Inchi Key MEBWABJHRAYGFW-DUXPYHPUSA-N

Description

Synonym (2E)-3-(2,4-Dichlorophenyl)prop-2-enoic acid, (E)-3-(2,4-Dichlorophenyl)acrylic acid
Purity 98%
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 233-235
Product Link https://store.apolloscientific.co.uk/product/trans-24-dichlorocinnamic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/trans-24-dichlorocinnamic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart