APOH0430C0CA Apollo Scientific Ltd

2-Nitrochalcone

  • MDL number MFCD00031069
  • Cas Number : 7473-93-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 124-128
Synonym 3-(2-Nitrophenyl)-1-phenylprop-2-en-1-one
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16036

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16036
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart