APOH0430C0D1 Apollo Scientific Ltd

Ethyl 4-(diethoxyphosphoryl)-3-methylcrotonate

  • MDL number MFCD08445507
  • Cas Number : 41891-54-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Ethyl 4-(diethoxyphosphoryl)-3-methylbut-2-enoate, Diethyl [3-(ethoxycarbonyl)-2-methylprop-2-en-1-yl]phosphonate
Density 1.089
Flash Point 113
Storage temp. Ambient temperature
Physical Form Liquid-Clear
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 284-287
Product Link http://www.apolloscientific.co.uk/display_item.php?id=16951

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=16951
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart